Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol 400 Monostearate, Spectrum™ Chemical
SDP

Catalog No. 18605327 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
18 kg
200 kg
Packaging:
Amber Glass Bottle
Poly Drum
Poly-Coated Glass Bottle

Catalog No. 18-605-327

Supplier: Spectrum Chemical Manufacturing Corporation P1154500GM

Only null left
Add to Cart
Add to Cart

P1154, 9004-99-3

Spectrum™ Chemical Polyethylene Glycol 400 Monostearate , commonly referred to as PEG 400 MONOSTEARATE, is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9004-99-3
Molecular Formula C20H40O3
Molecular Weight (g/mol) 328.54
InChI Key RFVNOJDQRGSOEL-UHFFFAOYSA-N
IUPAC Name 2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCO

Specifications

Acid Value 2
CAS Max % 1
Quantity 500 g
Grade Reagent
Hydroxyl Value 80-100
Moisture 0.03
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.