Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyethylene Glycol 400, NF, Spectrum™ Chemical

Click to view available options
Quantity:
25 mL
500 mL
2.5 L
20 L
Packaging:
Amber Glass Bottle
Glass Bottle
Poly Drum
Description
Spectrum™ Chemical Polyethylene Glycol 400, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
pH | 4.5 to 7.5 |
CAS Max % | 1 |
Quantity | 2.5 L |
Grade | NF |
Ignition Residue | 0.001 |
Packaging | Glass Bottle |
Viscosity | 6.8-8.0 cSt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction