Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyethylene Glycol 8000, Crystalline Powder, BAKER™, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
1 kg
2 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Specifications
Specifications
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Formula Weight | 7000 to 9000 (Average) |
Packaging | Wide Mouth Amber Glass Bottle |
Chemical Name or Material | Polyethylene Glycol 8000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction