Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyethylene glycol diacrylate, M.W. 1,000
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
PEGDA is used in dental compounds, offset inks, varnishes and flexible coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Granular Solid |
Solubility Information | Soluble in THF and chloroform. |
Quantity | 5 g |
Sensitivity | Light sensitive |
Chemical Name or Material | Polyethylene glycol diacrylate, M.W. 1,000 |
Safety and Handling
Hazard Category | H315-H317-H318-H335 |
Hazard Statement | GHS H Statement H318-H317-H335-H315 Causes serious eye damage. May cause an allergic skin reaction. May cause respiratory irritation. Causes skin irritation. |
Precautionary Statement | P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c |
EINECSNumber | 607-960-5 |
TSCA | Yes |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction