Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene glycol diacrylate, M.W. 3,400

Catalog No. p-7080129
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 26570-48-9 | C5H10O4 | 134.131 g/mol

Polyethylene glycol diacrylate is used in dental compounds, offset inks, varnishes and flexible coatings. It widely used in various biomedical applications such as hydrogel development for drug delivery, tissue engineering, wound healing and electrospinning processes for bioapplication due to its non-toxicity and cytocompatiblity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26570-48-9
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.131
MDL Number MFCD00081876
InChI Key FCDZZFLRLVQGEH-UHFFFAOYSA-N
Synonym acrylic acid; ethylene glycol, poly ethylene glycol diacrylate, polyethylene glycol diacrylate, poly ethylene glycol acrylate, average mn ∼375
PubChem CID 16212859
IUPAC Name ethane-1,2-diol;prop-2-enoic acid
SMILES C=CC(=O)O.C(CO)O

Specifications

Color White
Physical Form Granular Solid
Solubility Information Miscible with water. Soluble in THF,Chloroform.
Quantity 250 mg
Sensitivity Light sensitive
Chemical Name or Material Polyethylene glycol diacrylate, M.W. 3,400
Hazard Category H315-H317-H318-H335
Hazard Statement GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
EINECSNumber 607-960-5
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.