Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene glycol dimethacrylate, M.W. 8,000

Catalog No. AA4698503
Click to view available options
Quantity:
1 g
5 g

CAS: 25852-47-5 | CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3

Polyethylene glycol dimethacrylate is used as adhesives, coatings, sealants, photo resists, solder masks and photopolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25852-47-5
MDL Number MFCD00081877
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate

Specifications

Color White
Physical Form Granular Solid
Molecular Formula CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Solubility Information Soluble in water.
Quantity 1 g
Sensitivity Light and moisture sensitive
Chemical Name or Material Polyethylene glycol dimethacrylate, M.W. 8,000
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.