Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene glycol methyl ether methacrylate, M.W. 1,000, Thermo Scientific™

Catalog No. AA4699106
Click to view available options
Quantity:
250 mg
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26915-72-0
Molecular Formula C13H24O6
Molecular Weight (g/mol) 276.329
MDL Number MFCD00241432
InChI Key KRCGBOKYIUDIFY-UHFFFAOYSA-N
Synonym 2-methoxyethyl methacrylate, beta-methoxyethyl methacrylate, methacrylic acid, 2-methoxyethyl ester, 2-propenoic acid, 2-methyl-, 2-methoxyethyl ester, poly ethylene glycol methyl ether methacrylate, pegmem, peg-mem, acmc-209oay, methoxyethyl methacrylate, 2-methoxyethylmethacrylate
PubChem CID 93675
IUPAC Name 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOCCOCCOCCOC

Specifications

Color White
Physical Form Viscous liquid
Melting Point 33°C to 38°C
Solubility Information Soluble in water.
Quantity 5 g
Chemical Name or Material Polyethylene glycol methyl ether methacrylate, M.W. 1,000
Hazard Category H315-H317-H318-H335
Hazard Statement GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
TSCA Yes
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.