Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol, MP Biomedicals™

Catalog No. MP021519165 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021519165 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021519165 Supplier MP Biomedicals, Inc Supplier No. 02151916.5
Add to Cart
Add to Cart

Formed as a condensation polymer of ethylene oxide and water. Polyethylene Glycol, MP Biomedicals (PEG) has been used in many applications from medicine to industrial manufacturing, including the precipitation of proteins.

PEG has been used in a wide variety of biomedical and laboratory applications, including:

  • The precipitation of proteins
  • As a fusing agent in enhancing the effect of macrophages on hybridoma
  • In the separation and purification of biomolecules
  • In induction of cell hybridization
  • To recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples
  • Demonstrated to have anticell proliferation and antitumor effects via induction of apoptosis

Utilization of PEG:

  • PEGs are susceptible to oxidative degradation in the presence of air; minimizing exposure to elevated temperatures and oxygen or addition of an antioxidant can limit the amount of degradation
  • PEGs do not hydrolyze or deteriorate upon storage
  • PEGs do not support the growth of molds
  • PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, polyethylene glycol, macrogol
PubChem CID 174
ChEBI CHEBI:30742

Specifications

Melting Point 55°C to 65°C
Density 1.1–1.2g/mL (Lit.)
pH 4.5 to 7.5 (5% aq. soln.)
Quantity 500 g
Solubility Information Soluble in aqueous solution (100mg/mL, clear and colorless solution).
Vapor Pressure <0.01mmHg at 20°C (Lit.)
Formula Weight 3,400 average
Physical Form Powder
Recommended Storage Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.