Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene oxide, M.W. 300,000

Catalog No. AA4223722
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene oxide, M.W. 300,000 is used to impart viscosity to and modify flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
pH 5 to 7
Physical Form Powder
Melting Point 65°C
Density 1.21 g/mL
Solubility Information Soluble in acetone,alcohol,chloroform,toluene and dichloromethane.
Quantity 100 g
Odor Odorless
Refractive Index 1.4539
Chemical Name or Material Polyethylene oxide, M.W. 300,000
EINECSNumber 500-038-2
RTECSNumber TQ3630000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.