Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene rod, High Density, 19mm (0.75 in.) dia. 2.5cm, Thermo Scientific™

Catalog No. p-7052205
Click to view available options
Quantity:
2.5 cm
30 cm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-88-4
Molecular Formula (C2H4)n
Molecular Weight (g/mol) 28.05
MDL Number MFCD00084423
InChI Key VGGSQFUCUMXWEO-UHFFFAOYSA-N
Synonym ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
PubChem CID 6325
ChEBI CHEBI:18153
IUPAC Name ethene
SMILES *-CC-*

Specifications

Physical Form Rod
Quantity 2.5 cm
Odor Odorless
Chemical Name or Material Polyethylene rod, High Density, 19mm (0.75 in.) dia.
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.