Learn More
Valutek™ Standard Weight Polyethylene Shoe Cover
Catalog No. p-7162568
Click to view available options
Color:
Blue
White
This item is not returnable. View return policy
This item is not returnable. View return policy
Manufactured with 100% cross-linked polyethylene with heat-sealed seams. Strong, durable, impervious to non-hazardous liquids, and critical environment compatible.
  • All Valutek™ shoe covers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and stringent process control to ensure Valutek™ quality standard and product specification compliance
  • This product is recommended for use in a cleanroom Class 100 -1,000 (ISO 5-6) critical environment
  • Provides full liquid protection and is commonly used in microelectronics and pharmaceutical industries
Specifications
Material Polyethylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Seam Type Heat-sealed
Packaging Quantity 300/Cs.
Packaging Type 100/Bag, 3 Bags/Cs.
Length (English) 16 in.
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100-1000 (ISO 5-6)
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.