Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Polygodial

Catalog No. 319710
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 31-971-0

Supplier: Tocris Bioscience™ 3197/10

This item is not returnable. View return policy

TRPA1 channel activator; analgesic and antifungal

TRPA1 channel activator (EC50 = 400 nM) that is selective against TRPV1, TRPV2, TRPV3, TRPV4 and TRPM8 channels. Acts as an analgesic via desensitization of sensory neurons. Also exhibits antifungal activity via inhibition of mitochondrial ATPase.

Chemical Identifiers

CAS 6754-20-7
Molecular Formula C15H22O2

Specifications

Quantity 10 mg
Formula Weight 234.33
Purity >97%
Molecular Weight (g/mol) 234.34
Chemical Name or Material Polygodial
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.