Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyimide film, 0.013mm thick, Thermo Scientific Chemicals

Catalog No. p-7051745
Click to view available options
Quantity:
100 x 100 mm
150 x 150 mm
300 x 300 mm

Polyimide film is used in electronic and integrated circuit industries. It is also used for the reverse osmotic film in purification of water and the concentration of dilute materials from water. It finds application in making high temperature tape, IC Passivation coating, LCD alignment layer and insulated materials. It plays an important role in consumer electronic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62929-02-6
Molecular Formula C35H28N2O7
Molecular Weight (g/mol) 588.62
MDL Number MFCD00677744
InChI Key CDTDIQLZIBORMV-UHFFFAOYNA-N
Synonym polyimide resin, polyimide resin 10g, 1,3-isobenzofurandione, 5,5'-carbonylbis-, polymer with 1 or 3-4-aminophenyl-2,3-dihydro-1,3,3 or 1,1,3-trimethyl-1h-inden-5-amine, 1-4-aminophenyl-1,3,3-trimethyl-2h-inden-5-amine; 5-1,3-dioxo-2-benzofuran-5-carbonyl-2-benzofuran-1,3-dione
PubChem CID 6454485
SMILES CC1(C)CC(C)(C2=CC=C(N)C=C12)C1=CC=C(N)C=C1.O=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1

Specifications

Physical Form ≈0.1846g/100x100mm
Solubility Information Insoluble in water.
Quantity 100 x 100 mm
IUPAC Name 1-(4-aminophenyl)-1,3,3-trimethyl-2,3-dihydro-1H-inden-5-amine; 5-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-carbonyl)-1,3-dihydro-2-benzofuran-1,3-dione
Chemical Name or Material Polyimide film, 0.013mm thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.