Learn More
Polymedco™ Polymedco BTA stat™ Test Kit
SDP
Catalog No. p-5160374
Click to view available options
Includes:
10 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 2 Instructions for Home Use
30 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 5 Instructions for Home Use
Quantity:
10 Tests
30 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy
For detection of recurrent bladder cancer in five minutes
  •  Test uses monoclonal antibodies to detect presence of bladder tumor associated antigen in urine
  • Detects early stage and grade cancers that cytology often misses
  • Returns fast, objective result, a line in patient window indicates positive result
  • Requires only one voided urine sample, with no sample preparation
  • Adjunct test to cystoscopy
Specifications
Type BTA stat™ Test Kit
Includes 10 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 2 Instructions for Home Use
Quantity 10 Tests
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.