Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polymedco™ SED-CHEK 2™ ESR Controls
Shop All Polymedco Products

Click to view available options
Type:
Abnormal Control
Bi-Level Control
Normal Control
For Use With (Equipment):
For Sedimat and Manual Procedures
For use with Sedimat™ 15 automated ESR analyzer
Sedimat™ 15 automated ESR analyzer
Quantity:
3 x 8 mL
6 x 8 mL
Description
- Bi-level, whole-blood control designed to monitor ESR procedures
- Monitor technique as well as environmental and physical factors that adversely affect ESR results
- Run like a patient sample—not necessary to void diluent from pre-filled tubes
- Universal control assayed for both automated and manual ESR methods
Order Info
Controls packaged in 6 x 8mL vials
Specifications
Specifications
| Cap Type | Automatic self-zeroing |
| Compatible Methods | Both automated and manual ESR methods |
| Content And Storage | 20°C to 22°C |
| For Use With (Equipment) | For Sedimat and Manual Procedures |
| Type | Abnormal Control |
| Intended Use | Whole Blood reference control material designed to monitor ESR procedures |
| Packaging | Vial |
| Quantity | 6 x 8 mL |
| Stability | 31 Days Open Vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction