Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco™ Venoplast™ Vacuum and Nonvacuum ESR Tubes
SDP

Catalog No. p-4653648
Click to view available options
Product Type:
ESR Non-vacuum Tube no Diluent
ESR Non-vacuum Tube w/ Diluent
ESR Vacuum Tube with Diluent
Quantity:
1000 Box
200 Box

For use with Polymedco Sediten 10-Minute ESR Reader

Available vacuum or nonvacuum and with or without diluent.

Specifications

Product Type ESR Vacuum Tube with Diluent
For Use With (Equipment) Polymedco Sediten 10-Minute ESR Reader
Quantity 200 Box
Includes Diluent
Certifications/Compliance US Federal and state regulations
Media Type 3.8% Sodium Citrate
pH 5.65
Format Clear Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.