Learn More
PTS Diagnostics A1CNow+™ Systems
Catalog No. p-4830362
Click to view available options
Quantity:
10-Count Test Kit
20-Count Test Kit
This item is not returnable. View return policy
Catalog No. 21-124-414
Supplier: PTS Diagnostics 3021
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Provides fast and easy way of obtaining accurate A1C results
  • Results in five minutes
  • Requires only small volume (5μL) of blood sample
  • CLIA waived
  • Portable and can be used in multiple exam rooms
  • Easy to use and hence requires minimal training
  • Maintenance-free
  • Reimbursable
Certifications
National Glycohemoglobin Standardization Program (NGSP), IFCC
Specifications
Sample Type Blood
Quantity 20-Count Test Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.