Learn More
Cytiva Polymeric Manifold Tray for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
Catalog No. 45002622 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Ettan IPGphor 3 IEF Unit
Catalog No. For Use With (Equipment)
45-002-622 Ettan IPGphor 3 IEF Unit
1 options
Catalog No. 45-002-622 Supplier Cytiva Supplier No. 11002580
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Polymeric Manifold Tray for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
  • Cup for loading
  • For use with Ettan IPGphor 3 isoelectric focusing unit
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ettan IPGphor 3 IEF Unit
Type Ettan IPGphor Cup Loading Manifold Polymeric Tray
Material Plastic
No. of Gels 1 to 12
Quantity Complete packsize