Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymethylhydrosiloxane, ACROS Organics™

Click to view available options
Quantity:
100g
250g
500g
Packaging:
Glass bottle
9004-73-3
C5H11OSi
115.227
MFCD00084478
TXYXVOOPRILIHR-UHFFFAOYSA-N
polymethyl hydrosiloxane, 2-methyl-1,2-oxasilinanato, ch3 3sio ch3 hsio nsi ch3 3
12487658
2-methyl-1,2$l^{3}-oxasilinane
C[Si]1CCCCO1
9004-73-3
C5H11OSi
115.227
MFCD00084478
TXYXVOOPRILIHR-UHFFFAOYSA-N
polymethyl hydrosiloxane, 2-methyl-1,2-oxasilinanato, ch3 3sio ch3 hsio nsi ch3 3
12487658
2-methyl-1,2$l^{3}-oxasilinane
C[Si]1CCCCO1
Reduction reagent
Density 1.0000g/mL
Linear Formula (CH3)3SiO[(CH3)HSiO]nSi(CH3)3
Physical Form Viscous Liquid
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3970 to 1.4010
Solubility Solubility in water: practically insoluble. Other solubilities: may react, soluble in most common organic solvents, (should not be used in halocarbon solvent because, of the possibility of hydride-halide interchange, on catalysis or with heating)
Specific Gravity 1
Viscosity 15-30 mm2/s (25°C)
Color Colorless
Quantity 500g
Flash Point 121°C
Chemical Name or Material Polymethylhydrosiloxane
Show More Show Less