Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Polymyxin B Monoclonal Antibody (45)
GREENER_CHOICE

Catalog No. 01676524
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-524 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-524 Supplier Invitrogen™ Supplier No. MA140133
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody binds to free Polymyxin B as well as to Polymyxin B already bound to LPS.

The peptide antibiotic Polymyxin B (PMB) binds to bacterial endotoxin (lipopolysaccharide, LPS). The interaction of PMB with LPS involves ionic forces between amino groups in PMB and negatively charged phosphate and carboxyl groups in the lipid A-Kdo region. PMB has relevance for endotoxin research in at least two ways: first, PMB reacts with LPS of many species regardless of varied serospecificity, and thus it can be used as a general probe for measuring or detecting LPS or lipid A. Second, binding of PMB to LPS may result in neutralization of the detrimental effects of LPS either in vitro or in vivo. Antibodies to PMB allow for the quantitative study of interaction of PMB/LPS interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polymyxin B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 45
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.02% sodium azide
Gene Alias PMB
Host Species Mouse
Immunogen Polymixin B.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.