Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polymyxin B Sulfate, Micronized, USP, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
5 gal.
Packaging:
Fiber Drum
Description
Spectrum™ Chemical Polymyxin B Sulfate, Micronized, USP is an antibiotic that is primarily used to treat resistant Gram-negative infections. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS | 1405-20-5 |
CAS Max % | 1 |
Ignition Residue | 0.05 |
Loss on Drying | 0.07 |
Packaging | Fiber Drum |
Quantity | 5 gal. |
Molecular Formula | C48H84N16O17S |
InChI Key | HNDFYNOVSOOGDU-UHFFFAOYNA-N |
SMILES | OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O |
IUPAC Name | 4-amino-N-{1-[(3-amino-1-{[6,9,18-tris(2-aminoethyl)-15-benzyl-3-(2-hydroxyethyl)-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl)carbamoyl]-3-hydroxypropyl}-2-formamidobutanamide; sulfuric acid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction