Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polynuclear Aromatic Hydrocarbons Antibody (BAP-13), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500479V
Description
Polynuclear Aromatic Hydrocarbons Monoclonal specifically detects Polynuclear Aromatic Hydrocarbons in Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Polynuclear Aromatic Hydrocarbons | |
Monoclonal | |
DyLight 405 | |
PAHs | |
Benzo[a]pyrenyl-1-butyric acid conjugated to BSA. | |
0.1 mL | |
Primary | |
Rat | |
IgG1 |
ELISA, Immunocytochemistry, Immunofluorescence | |
BAP-13 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Affinity Purified | |
RUO | |
The antibody BAP-13 is specific for Polynuclear Aromatic Hydrocarbons. The antibody BAP-13 inhibits DNA adduct formation (this ability was investigated in rat liver microsomes spiked with calf thymus DNA and 7,8-diol-B[a]P). | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction