Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyoma Virus Early Antigen Antibody (PyC), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB1002793F
Description
Polyoma Virus Early Antigen Monoclonal specifically detects Polyoma Virus Early Antigen in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Polyoma Virus Early Antigen | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Rat | |
Purified |
Immunocytochemistry, Immunofluorescence | |
PyC | |
Immunocytochemistry/Immunofluorescence | |
Large T antigen, Middle T antigen, Polyoma virus large T antigen, Polyoma virus middle T antigen, Polyoma virus small T antigen, Polyomavirus early antigen, Polyomavirus large T antigen, Polyomavirus middle t antigen, Polyomavirus small T antigen, Small T antigen | |
Polyoma virus-transformed Wistar rat fibroblast cellline REWA5/T1A1. | |
0.1 mL | |
Virology Bacteria and Parasites | |
Allows the detection and isolation of all three viral T antigens. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction