Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyoma Virus, Large T Antigen Antibody (PyLT), Alexa Fluor™ 750, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB1002755AF750
Description
Polyoma Virus, Large T Antigen Monoclonal specifically detects Polyoma Virus, Large T Antigen in Virus samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Polyoma Virus, Large T Antigen | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Polyoma virus-transformed Wistar rat fibroblast cell line Py REWA5/T1A1. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
PyLT | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Rat | |
Protein G purified | |
Virology Bacteria and Parasites | |
Virus | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction