Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Polyoma Virus Large T Monoclonal Antibody (PyLT)
GREENER_CHOICE

Catalog No. MA146063
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA146063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA146063 Supplier Invitrogen™ Supplier No. MA146063
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Polyoma Virus Large T Monoclonal Antibody for Western Blot, ICC/IF, IP

The mouse polyoma virus (Py) is an oncogenic virus, which encodes for three early proteins, large, middle and small T (tumour) antigen. PyLT allows isolation of viral T antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polyoma Virus Large T
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PyLT
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias Jvgp5; PyLT
Host Species Rat
Immunogen Polyoma virus-transformed Wistar rat fibroblast cell line Py REWA5/T1A1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.