Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyoma Virus, Medium T Antigen Antibody (PyMT), Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $501.50

Specifications

Antigen Polyoma Virus, Medium T Antigen
Clone PyMT
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 19816927), Dual RNAscope ISH-IHC reported in scientific literature (PMID 30367629)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002749SS
SDP
View Documents
Novus Biologicals
NB1002749SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB1002749
SDP
View Documents
Novus Biologicals
NB1002749
0.1 mL
Each for $501.50
Only null left
Add to Cart
 
Description

Description

Polyoma Virus, Medium T Antigen Monoclonal specifically detects Polyoma Virus, Medium T Antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.
Specifications

Specifications

Polyoma Virus, Medium T Antigen
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rat
Virology Bacteria and Parasites
PBS with 0.02% Sodium Azide
Polyoma Virus-transformed Wistar rat fibroblast cell line Py REWA5/T1A1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
PyMT
Western Blot 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 19816927), Dual RNAscope ISH-IHC reported in scientific literature (PMID 30367629)
Monoclonal
Purified
RUO
Virus
Middle t antigen, MPyVgp2, Polyoma virus middle T antigen, Polyomavirus medium t antigen, Polyomavirus middle t antigen
IgG2b
Protein G purified
Polyoma Virus.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.