Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyphenylmethylsiloxane, MW 2500-2700

Catalog No. p-7059377
Click to view available options
Quantity:
50 g
250 g
1 kg

CAS: 9005-12-3 | C7H8OSi | 136.23 g/mol

A biochemical for proteomics research. A novel coating for solid-phase microextraction. A polyphenylmethylsiloxane for use in skincare, suncare, color cosmetic, hair care, and antiperspirant / deodorant applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-12-3
Molecular Weight (g/mol) 136.23
MDL Number MFCD00146731

Specifications

Physical Form Liquid
Molecular Formula C7H8OSi
Synonym methylphenylsilanone, polyphenylmethylsiloxane, methylphenyl oxo silane, silane, methyloxophenyl, methyl phenyl silanone
Solubility Information Insoluble in water.
Quantity 50 g
InChI Key IXKNJYSAWNJQQY-UHFFFAOYSA-N
SMILES C[Si](=O)C1=CC=CC=C1
IUPAC Name methyl-oxo-phenylsilane
PubChem CID 3032548
Flash Point 300°C (572°F)
Chemical Name or Material Polyphenylmethylsiloxane, M.W. 2,500 to 2,700
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.