Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Polypropylene Conical Flasks with Screw Caps
SCI_ED

Catalog No. S13966 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S13966 250 mL
S89384 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S13966 Supplier Eisco™ Supplier No. CH0431B
Add to Cart
Add to Cart

Eisco™ Polypropylene Conical Flasks with Screw Caps feature a rigid, durable, and translucent body.

  • Plastic conical flask with screw cap
  • Durable polypropylene construction
  • Autoclavable

Specifications

Closure Color Clear
Closure Material Polypropylene
Color Clear
Capacity (Metric) 250 mL
Material Polypropylene
For Use With (Application) Heating and holding liquids
Product Type Conical Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.