Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Polypropylene Cryo Cube Box
SDPSCI_ED

Catalog No. S01151 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Holds:
100 Tubes
81 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
S01151 81 Tubes
S01152 100 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S01151 Supplier United Scientific™ Supplier No. 66501
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Storage boxes that can be used with multiple tube sizes.

  • Manufactured of polypropylene, these autoclavable boxes are stackable for sample storage in 1.5 to 2.0 mL microtubes and 1.0 to 2.0 mL cryogenic vials
  • Boxes can be used at temperatures from -90°C to +121°C and are autoclavable
  • Boxes feature a hinged lid connected to the base and a locking system to ensure secure closure
  • A molded alphanumeric grid allows for easy identification of samples
  • Dimensions of cryo boxes are designed to be compatible with vertical, horizontal, and drawers style freezer racks
  • Each pack includes two yellow boxes and two blue boxes

Specifications

Closure Material Polypropylene
Color Blue
Material Polypropylene
Shape Square
Type Cryo Box
Holds 81 Tubes
Dimensions (L x W x H) 128 x 128 x 54 mm

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.