Learn More
Valutek™ Polypropylene Fabric Grounding Ribbon Shoe Cover, 16 in.
Catalog No. p-7162571
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Manufactured with 100% non-linting, non-woven spunbond polypropylene fabric. This shoe cover has stitched seams, sprayed latex anti-skid treads on the bottom for safety
  • 19.6 in. long conductive ribbon stitched on a solid seam to provide ESD grounding
  • Lightweight, strong, and durable.
  • All Valutek™ shoe covers are tested and manufactured in ISO compliant facilities under individual inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environment
  • It is commonly used for protecting against dirt, grime and certain dry particulates in nonhazardous environments and for protecting sensitive devices in the work area
Specifications
Size Large
Material Polypropylene
Sterility Non-sterile
Sole Type Anti-skid
Description Disposable anti-skid SPP ESD shoe cover
Packaging Quantity 300/Cs.
Packaging Type 100/Bag, 3 Bags/Cs.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.