Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polypropylene sheet, 6.36mm (0.25in) thick

Catalog No. aa45174hb
Click to view available options
Quantity:
300 x 300 mm

CAS: 9003-07-0 | (C3H6)n | 354.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-07-0
Molecular Formula (C3H6)n
Molecular Weight (g/mol) 354.58
MDL Number MFCD00084447
InChI Key NSYDMBURIUSUDH-UHFFFAOYNA-N
Synonym oxiranedodecanoic acid, 3-octyl-, cis, 12-2s,3r-3-octyloxiran-2-yl dodecanoic acid, polypropylene, 2-oxiranedodecanoic acid, 3-octyl-, 2r,3s-rel, oxiranedodecanoic acid, 3-octyl-, 2r,3s-rel, 3beta-octyloxirane-2beta-dodecanoic acid, 12-3-octyl-2-oxiranyl dodecanoic acid #, special for nanometer polypropylene pipe material
PubChem CID 76958
IUPAC Name 12-[(2S,3R)-3-octyloxiran-2-yl]dodecanoic acid
SMILES CC(-*)C-*

Specifications

Quantity 300 x 300 mm
Chemical Name or Material Polypropylene sheet, 6.36mm (0.25 in.) thick
RTECSNumber UD1842000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.