Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Polyprotein P1234 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144583
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144583 Supplier Invitrogen™ Supplier No. PA5144583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Polyprotein P1234 Polyclonal Antibody for Western Blot, ELISA

[Polyprotein P1234]: Inactive precursor of the viral replicase, which is activated by cleavages carried out by the viral protease nsP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Polyprotein P1234
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.664 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Non-structural polyprotein
Host Species Rabbit
Immunogen Recombinant Venezuelan equine encephalitis virus Polyprotein P1234 protein (1887-2002aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.