Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

polyQ specific Mouse anti-Rat, Mouse, Human, Unconjugated, Clone: MW1, MilliporeSigma™

Catalog No. MABN242725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2427-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2427-25 Supplier MilliporeSigma Supplier No. MABN242725UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-polyQ specific, clone MW1, Cat. No. MABN2427, is a highly specific mouse monoclonal antibody that targets expanded PolyQ repeats from Huntingtin (Htt) and has been tested for use in Immunocytochemistry, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen polyQ specific
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MW1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Htt mutant;PolyQ-Htt;Clone MW1;Atrophin-1
Host Species Mouse
Immunogen GST-tagged recombinant proteins expressed from two constructs containing the polyQ domain (19 repeats) and 34 amino acids of the dentatorubralpalliodoluysian atrophy (DRPLA) gene.
Molecular Weight of Antigen ∽350 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone MW1 specifically recognize the polyQ domain and does not react with wild-type Htt in any significant manner.
Target Species Rat, Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.