Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polysorbate 40, NF, Spectrum™ Chemical

Click to view available options
Quantity:
25 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Description
Spectrum™ Chemical Polysorbate 40, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
Acid Value | 2 |
CAS Max % | 1 |
Quantity | 500 mL |
Hydroxyl Value | 89-105 |
Ignition Residue | 0.0025 |
Packaging | Amber Glass Bottle |
Saponification Value | 41-52 |
Water | 0.03 |
Grade | NF |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction