Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 60

Catalog No. AAL15031AE
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. AAL15031AE

Supplier: Thermo Scientific Chemicals L15031AE

Only null left
Add to Cart

CAS: 9005-67-8 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | 522.68 g/mol

Polysorbate 60 is used in food, medicine, plastic, cosmetics and other industries. It is an excellent emulgator and one of elements to constitute polyacrylonitrile fiber spinning oil. It also used as softening agent after processing fiber and eliminate fiber static and improve its softness.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-67-8
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD01780110
InChI Key DEKFZWMENCCOFU-UHFFFAOYNA-N
Synonym Polyoxyethylene sorbitan monostearate
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Density 1.08
Merck Index 14,7582
Solubility Information Soluble in water and alcohols. Insoluble in oils.
Quantity 100 mL
Flash Point >150°C (302°F)
Chemical Name or Material Polysorbate 60
EINECSNumber 000-000-0
RTECSNumber WG2934000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.