Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 80, FCC, 65-69.5%, Spectrum™ Chemical
SDP

Catalog No. 18605391
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum

P1179, 9005-65-6

Spectrum™ Chemical Polysorbate 80, FCC is used as a wetting agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Acid Value 2
CAS Max % 1
Quantity 20 L
Hydroxyl Value 65 -80
Ignition Residue 0.0025
Packaging Poly Drum
Saponification Value 45-55
Water 0.03
Percent Purity 65 to 69.5%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.