Learn More
Polystyrene, atactic
Description
Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Beads |
Solubility Information | Insoluble in water. |
Quantity | 50 g |
IUPAC Name | styrene |
Formula Weight | 125,000-250,000 |
Odor | Odorless |
Chemical Name or Material | Polystyrene, Atactic |
Safety and Handling
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.