Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polystyrene for GPC 1,000, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Physical Form | Neat |
Linear Formula | [CH2 CH(C6H5)]n |
MDL Number | MFCD00084450 |
Synonym | PS |
Type | Analytical Standard |
Quantity | 250 mg |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction