Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polystyrene for GPC 30,000, MilliporeSigma™ Supelco™

Catalog No. p-7187529 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 mg
1 g

Specifications

Physical Form Neat
Linear Formula [CH2 CH(C6H5)]n
MDL Number MFCD00084450
Synonym PS
Type Analytical Standard
Quantity 250 mg
Grade GPC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.