Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polystyrene latex microsphere, 90.0 micron, 2.5 wt% dispersion in water

Catalog No. aa42751aa
Click to view available options
Quantity:
5 mL

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol

Polystyrene latex microsphere is used as a biocompatible material, especially in dentistry. They are used as insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads. It is also used as extrusion molded foamed packaging, dinnerware, bottles, toys and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-53-6
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name styrene
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Color White
Physical Form Dispersion
Solubility Information Fully miscible with water.
Quantity 5 mL
Concentration or Composition (by Analyte or Components) 2.5 wt% dispersion in water
Odor Odorless
Particle Size 90.0 Micron
Chemical Name or Material Polystyrene latex microsphere
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.