Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polystyrene Standards, Low Molecular Weight, Analytical Standard, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Physical Form | Neat |
Quantity | 1 Kit |
Grade | Analytical Standard |
Concentration or Composition (by Analyte or Components) | MW 2500 250 mg; MW 5000 250 mg; MW 9000 250 mg; MW 17,500 250 mg; MW 30,000 250 mg; MW 50,000 250 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction