Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysulfone Resin, Pellets

Catalog No. AC178912500
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass Bottle
Plastic bottle

CAS: 25135-51-7 | (C27H22O4S)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25135-51-7
Molecular Formula (C27H22O4S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00134387
InChI Key ZUZZUJNBGCVPLQ-UHFFFAOYSA-N
PubChem CID 16211415
SMILES CC(C)(C1=CC=C(O-*)C=C1)C1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(-*)C=C2)C=C1

Specifications

Color Amber to Beige
Physical Form Pellets
Density 1.2400g/mL
Solubility Information Solubility in water: insoluble.
Quantity 250 g
IUPAC Name 4-(4-hydroxybenzenesulfonyl)phenol; 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Plastic bottle
Refractive Index 1.633
Specific Gravity 1.24
Chemical Name or Material Polysulfone resin, approx. M.W. 60,000
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.