Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinyl alcohol, 87-89% hydrolyzed, high molecular weight

Catalog No. AA4124022
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Physical Form Solid
Solubility Information Soluble in water.
Quantity 100 g
Percent Purity 87 to 89%
Odor Odorless
Chemical Name or Material Polyvinyl alcohol, High molecular weight, Hydrolyzed
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.