Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS Polyvinylidene Fluoride Sheet

Catalog No. 09924064 Shop All Osmonics Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Dimensions (LxW):
100 x 100 mm
150 x 150 mm
200 x 200 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Dimensions (LxW)
09-924-064 0.45 μm 100 x 100 mm
09-924-042 0.22 μm 200 x 200 mm
09-924-065 0.45 μm 200 x 200 mm
09-924-066 0.45 μm 150 x 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-924-064 Supplier GVS Supplier No. 1212644
Only null left
Add to Cart
Add to Cart

Naturally hydrophobic, unsupported transfer membrane. It has a high binding capacity, which prevents protein from passing through the membrane, and a low background that provides for an excellent signal-noise ratio.

Properties

  • This membrane also has broad chemical compatibility, which is important when used with common stains such as Amido Black, Colloidal Gold, Coomassie Blue, India Ink and Ponceau-S.
  • This Polyvinylidene fluoride membrane will not degrade, distort or shrink when a high concentration of methanol is used for destaining. Its exceptional strength, high binding capacity and chemical compatibility make GVS Polyvinylidene fluoride membrane ideal for use in Western blots, immunoblotting, and solid phase assays and plaque lifts.

Characteristics

  • Superior strength can withstand aggressive handling or be used with automated equipment without breaking or tearing
  • Low extractables ensures tests will be clean with consistent results
  • Exceptional sensitivity detects low-level components
  • Hydrophobic for high protein binding
  • Lot-to-lot consistency quality checks ensure consistent binding for dependable results every time
  • BSA protein binding capacity 125μg/cm²

Applications

  • Western blots
  • Immunoblotting
  • Solid phase assays
  • Amino acid or protein analyses

Specifications

Length (Metric) 100 mm
Width (Metric) 100 mm
Format Sheet
Material PVDF
Pore Size 0.45 μm
Dimensions (LxW) 100 x 100 mm
Thickness 140 to 250 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.