Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinylpyrrolidone, Fisher BioReagents™

Catalog No. BP431500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP431500 500 g Poly Bottle
BP431-100 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP431500

Polyvinylpyrrolidone, >-95%, C6H9NO, CAS Number-9003-39-8, n-vinyl-2-pyrrolidinone, 2-pyrrolidinone, 1-ethenyl, n-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, 1-vinylpyrrolidone, 1-vinyl-2-pyrrolidinone, 1-vinyl-2-pyrrolidone, n-vinylpyrrolidone, n-vinylpyrrolidinone, vinylpyrrolidone, 500g

Polyvinylpyrrolidone, >-95%, C6H9NO, CAS Number-9003-39-8, n-vinyl-2-pyrrolidinone, 2-pyrrolidinone, 1-ethenyl, n-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, 1-vinylpyrrolidone, 1-vinyl-2-pyrrolidinone, 1-vinyl-2-pyrrolidone, n-vinylpyrrolidone, n-vinylpyrrolidinone, vinylpyrrolidone, 500g

Polyvinyl Pyrrolidone is a component of Denhardt's Reagent, commonly used in nucleic acid hybridization procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White
pH 3 to 7
Physical Form Solid
Melting Point 100°C
Ash 0.05% max.
Assay Percent Range ≥95 %
Merck Index 15, 7814
Quantity 500 g
Percent Purity ≥95%
Identification Pass Test
Packaging Poly Bottle
Assay (dry basis)≥95%
Chemical Name or Material Polyvinylpyrrolidone
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.