Learn More
Polyvinylpyrrolidone, M.W. 360,000
Description
A cryoprotectant, also useful in plant cell culture research Polyvinylpyrrolidone is used as an adhesive in glue sticks; an emulsifier and a disintegrant for solution polymerization; an additive to Doro's RNA extraction buffer; as a liquid-phase dispersion enhancing agent in diffusion-ordered spectroscopy (DOSY) NMR and as a thickening agent in tooth whitening gels. It finds use in personal care products like shampoos and toothpastes, in ink for inkjet printers as well as in contact lens solutions. It is used as a food additive and in the wine industry as a fining agent for white wine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Merck Index | 14,7697 |
| Solubility Information | Soluble in water. |
| Quantity | 500 g |
| Formula Weight | 360 kd |
| Chemical Name or Material | Polyvinylpyrrolidone |
Safety and Handling
| Hazard Category | H500 |
| RTECSNumber | TR8370000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.