Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000

Catalog No. p-4509633
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 26837-42-3 | (C2H3KO4S)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26837-42-3
Molecular Formula (C2H3KO4S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00081923
InChI Key WZAPMUSQALINQD-UHFFFAOYSA-M
Synonym pvsk, potassium ethenyl sulfate, potassium poly vinyl sulfate, polyvinyl sulfate potassium salt, sulfuric acid, monovinyl ester, polymers, potassium salt, sulfuric acid, monoethenyl ester, homopolymer, potassium salt, vinyl sulfate potassium salt, sulfuric acid vinyl=potassium salt
PubChem CID 5055241
SMILES [K]OS(=O)(=O)OC(-*)C-*

Specifications

Color Beige-Brown to White
Physical Form Crystalline Powder
Linear Formula (-CH2CH(OSO3K)-)n
Quantity 5 g
IUPAC Name potassium ethenyl sulfate
Percent Purity ≥21.2% (K)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Polyvinylsulfuric acid potassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.