Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ PolyWear™ Smooth Disposable Gowns

Catalog No. 19125555
missing translation for 'orderingAttributeHoverText'
Color:
Blue
White
Yellow
Size:
Medium
Large
X-Large
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Lightweight, single-layer, low-density polyethylene gown for clinical and laboratory settings.

Lightweight, single-layer, low-density polyethylene gown for clinical and laboratory settings

Heat-sealed seam is behind sleeves—minimizes chance of contamination passing through.

  • Full-length wraparound with open back tailored to eliminate excess material
  • Tear-away collar for quick and easy removal
  • Thumb loop on sleeve keeps cuffs under gloves, enhancing leak protection
  • Gowns are folded for quick shake-out and easy donning
  • Powder blue with smooth matte finish

Order Info

Packed individually or bulk in dispenser box.

Compliance

Complies with FDA regulations. Successfully tested for liquid and aerosol blood resistance under ASTM™ F 1670 Synthetic Blood Penetration Resistance and ASTM 1671 Bacteriophage Penetration Resistance.

Specifications

For Use With (Application) Clinical, Laboratory
Gender Unisex
Size Large
No. of Pockets None
Product Type General Purpose Gown
Material Polyethylene Film (Virgin)
Color White
Disposable Single-use
Sterility Non-sterile
Finish Smooth Matte
Model 10502
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.