Learn More
POM121 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | POM121 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0372
![]() ![]() |
Proteintech Group Inc
156451AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
15645-1-AP do well in identifying the nuclear envelope in immunofluorescence experiments.
POM121 (POM121 membrane glycoprotein), also known as POM121A (nuclear envelope pore membrane protein POM 121A), NUP121 (nucleoporin Nup121) or pore membrane protein of 121 kDa, is a 1,249 amino acid single-pass membrane protein that exists as three alternatively spiced isoforms and belongs to the POM121 family. Encoded by a gene that maps to human chromosome 7q11.23, POM121 is highly conserved and localizes to nucleus membrane and endoplasmic reticulum membrane. An essential component of the nuclear pore complex (NPC), POM121 is mandatory for nuclear envelope formation and may play a role in biogenesis of the NPC. POM121 associates with the central spoke ring complex and is involved in anchoring components of the pore complex to the pore membrane by way of its F-X-F-G repeat-containing domain. POM121 overexpression induces the formation of cytoplasmic annulate lamellae (AL).Specifications
POM121 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
P52591, Q96HA1 | |
113975, 9883 | |
POM121 Fusion Protein Ag8177 | |
Primary | |
-20°C | |
POM121 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0618, Nucleoporin Nup121, NUP121, POM121, POM121A | |
Pom121 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.