Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Pomalidomide 4'-alkylC5-amine
GREENER_CHOICESDP

Catalog No. 671625 Shop All Novus Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-162-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 67-162-5 Supplier R&D Systems™ Supplier No. 6716/25

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Pomalidomide-alkylC5-amine

Pomalidomide 4'-alkylC5-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC5 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

This product has been recently renamed. The previous name for this product was Pomalidomide - linker 4

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material 4-[(5-Aminopentyl)amino]-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione hydrochloride
CAS 2375194-03-7
Synonym Pomalidomide - linker 4
Quantity 25 mg
Product Line Tocris
Molecular Formula C18H22N4O4.HCl
Formula Weight 394.86
Product Type Ligand
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.